Clamps for conical standard ground joints, KECK™, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
286336109 286336203 286336306 286336409 286336503 286336606 286336709 286336803 286336906
271-7111EA 3.8 GBP
271-7111 271-7112 271-7113 271-7114 271-7115 271-7116 271-7107 271-7108 271-7117
Clamps for conical standard ground joints, KECK™, DURAN®
Clamps NS Joint Clamps
POM. These clips made of temperature resistant polyacetal (POM) are easy, safe and can be fitted quickly.
  • Good spring properties
  • Colour coded by size
  • No risk of scratching the glass
  • Will not corrode or bend out of shape