Adapters, bends
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05020001 05020002 05020003 05020004
271-2201EA 38.7 GBP
271-2201 LENZ05020002 201-1175 271-2202
Adapters, bends
Distillation Components Distillation Adapters
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 75°. With two standard ground cones.