Flasks, round bottom with four necks and standard ground joints, side necks parallel
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03344458 03344470 03344473 03344479 03345470 03345473 03345485 03344485 03344476 03345482 03345479 03345476 03345488
271-1538EA 239 GBP
271-1538 271-1544 271-1537 271-1536 271-1545 271-1546 LENZ03345485 LENZ03344485 LENZ03344476 LENZ03345482 LENZ03345479 LENZ03345476 LENZ03345488
Flasks, round bottom with four necks and standard ground joints, side necks parallel
Flasks Round Bottom Flasks
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certifications: DIN 12392