Potassium hydrogen sulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

27011.237EA 75.1 GBP
27011.237 27011.294 27011.460
Potassium hydrogen sulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Potassium hydrogen sulphate
Formula: KHSO₄
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695

Order Now

Specification Test Results

Assay Min. 99.0 %
Heavy metals (as Pb) Max. 5 ppm
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 5 ppm
PO4 (Phosphate) Max. 10 ppm
Al (Aluminium) Max. 20 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 5 ppm
Na (Sodium) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR