High Purity rNTPs, Cytiva
Supplier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
27-1006-03 27-1200-04 27-2000-04 27-2025-01 27-2056-01 27-2066-01 27-2076-01 27-2086-01 27-2056-62 27-2066-62 27-2076-62 27-2086-62
27-1006-03EA 285 GBP
27-1006-03 27-1200-04 27-2000-04 27-2025-01 27-2056-01 27-2066-01 27-2076-01 27-2086-01 27-2056-62 27-2066-62 27-2076-62 27-2086-62
High Purity rNTPs, Cytiva
Nucleic Acid Reagents Nucleotides
Easy to use, time saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
  • Each lot is tested for ribonuclease contamination to ensure success in the application
  • Supplied as crystalline solid or in purified water as 100 mM solution (pH 7,5)
  • Available either as individual products or in a convenient set containing ATP, CTP, GTP and UTP in separate tubes
Kit contents: Adenosine 5'-Triphosphate, 100 mM Solution (ATP); Cytidine 5'-Triphosphate, 100 mM Solution (CTP); Guanosine 5'-Triphosphate, 100 mM Solution (GTP); Uridine 5'-Triphosphate, 100 mM Solution (UTP)