Potassium iodide 99,0-100,5 Ph. Eur.

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Dipotassium diiodide, Kali iodide

26850.230EA 290 GBP
26850.230 26850.296 26850.365 26850.260 26850.460
Potassium iodide 99,0-100,5 Ph. Eur.
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 681 °C
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Assay (calculated on dried substance) 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
Alkalinity Passes test
IO3 (Iodate) Passes test
SO4 (Sulphate) Max. 150 ppm
S2O3 (Thiosulphate) Passes test
Heavy metals (as Pb) Max. 10 ppm
Fe (Iron) Max. 20 ppm
Loss on drying (105°C) Max. 1 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR