Potassium fluoride 99,0, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

26820.236EA 113 GBP
26820.236 26820.293
Potassium fluoride 99,0, AnalaR NORMAPUR® zA
Potassium fluoride
Formula: KF
MW: 58.1 g/mol
Boiling Pt: 1505
Melting Pt: 860
Density: 2.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1 III
Merck Index: 13,07714

Order Now

Specification Test Results

Assay Min. 99.0 %
Acidity Max. 0.025 meq/g
Alkalinity Max. 0.01 meq/g
Heavy metals (as Pb) Max. 10 ppm
Cl (Chloride) Max. 50 ppm
SiF6 (Hexafluorosilicate) Max. 0.05 %
SO4 (Sulphate) Max. 0.05 %
Cu (Copper) Max. 25 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 10 ppm
Zn (Zinc) Max. 25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR