Potassium hydroxide 85,0-100,5, pellets Ph. Eur.

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potash caustic, Caustic potash

26670.294EA 99.6 GBP
26670.294 26670.363 26670.465
Potassium hydroxide 85,0-100,5, pellets Ph. Eur.
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Assay (calculated as KOH) 85.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
pH (20°C; 0,01 %) Min. 10.5
Identification B (K) Passes test
Solution S1 Passes test
Solution S2 Passes test
Appearance of solution Passes test
CO3 (as K2CO3) Max. 2.0 %
Cl (Chloride) Max. 200 ppm
PO4 (Phosphate) Max. 100 ppm
SO4 (Sulphate) Max. 200 ppm
Fe (Iron) Max. 10 ppm
Na (Sodium) Max. 1.0 %
Heavy metals (as Pb) Max. 10 ppm
Residual solvents Passes test
Conforms to Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR