Potassium hydroxide 47% aqueous solution, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potash caustic, Caustic potash

26633.290EA 75.4 GBP
26633.290 26633.460
Potassium hydroxide 47% aqueous solution, AnalaR NORMAPUR® zA
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.475 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Assay (acidimetric) 46.0 - 48.0 %
Carbonate (as K2CO3) max. 1.0 %
Chloride (Cl) max. 0.004%
Phosphate (PO4) max. 0.0005%
Sulfate (SO4) max. 0.002%
Total nitrogen (N) max. 0.0003%
Heavy metals (as Pb) max. 0.0005%
Al (Aluminium) max. 0.001%
As (Arsenic) max. 0.0001%
Ca (Calcium) max. 0.0005%
Cu (Copper) max. 0.0005%
Fe (Iron) max. 0.0005%
Mg (Magnesium) max. 0.0005%
Ni (Nickel) max. 0.0005%
Pb (Lead) max. 0.0005%
Zn (Zinc) max. 0.0005%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR