n-Pentane ≥95%, TECHNICAL

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

26192.463EA 458 GBP
26192.463 26192.292 26192.554 26192.361
n-Pentane ≥95%, TECHNICAL
n-Pentane
Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 95 %
Boiling point 35 - 37 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR