Bovine desoxyribonuclease I (from Pancreas)

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

260913-10 260913-25
260913-10EA 276 GBP
260913-10 260913-25
Bovine desoxyribonuclease I (from Pancreas)
Enzymes

Order Now

SPECIFICATIONS

Enzyme name Desoxyribonuclease I
Environmentally Preferable
Enzyme synonyms DNase I
Source Pancreas
Species Bovine
CAS No. 9003-98-9
Storage conditions Freezer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR