Isobutyl acetate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

259830010 259830025 259830250 259830000
ACRO259830010EA 35.7 GBP
ACRO259830010 ACRO259830025 ACRO259830250 259830000.
Isobutyl acetate 98%
Isobutyl acetate
Formula: C₆H₁₂O₂
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.873 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008932
CAS Number: 110-19-0
EINECS: 203-745-1
UN: 1213
ADR: 3,II
Merck Index: 13,05147

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3910 (20°C, 589 nm)
Water ≤0.03 %
Density (20°C) 0.871 to 0.875 kg/l
GC ≥97.5 %
Color scale ≤10 APHA
Acidity (CH3COOH) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR