1-Naphtholbenzeine analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: alpha-Naphtholbenzein, p-Naphtholbenzein, α-Naphtholbenzein, p-Naphtholbenzeine, 4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one, 1-Naphtholbenzein, Bis(4-hydroxy-1-naphthyl)phenylmethanol, 4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanol, α-Naphtholbenzeine

25768.136EA 412 GBP
25768.136
1-Naphtholbenzeine analytical reagent
1-Naphtholbenzeine
Formula: C₂₇H₁₈O₂
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00078492
CAS Number: 145-50-6
EINECS: 205-656-3

Order Now

Specification Test Results

Suited for acid-base indicator Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR