1,2,4-Trichlorobenzene ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

256412-1L 256412-2L
256412-1LEA 189 GBP
256412-1L 256412-2L
1,2,4-Trichlorobenzene ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
1,2,4-Trichlorobenzene

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.

Formula: C₆H₃Cl₃
Boiling Pt: 213.5 °C
Melting Pt: 17 °C
Density: 1.454 g/cm³ (20 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1,III
Merck Index: 13,09704

Order Now

Specification Test Results

Appearance of the substance complying
Identity (IR) complying
Assay (GC) Min. 99.0 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.01
Absorbance at 375 nm Max. 0.03
Absorbance at 350 nm Max. 0.1
Absorbance at 310 nm Max. 0.50
Absorbance at 308 nm Max. 1.00
Explosion Limit 6.6 %, 150 °F
Refractive Index n20/D 1.571 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR