Magnesium nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

25135.298EA 75.6 GBP
25135.298 25135.466
Magnesium nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 89 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9
EINECS: 233-826-7
UN: 1474
ADR: 5.1,III
Merck Index: 13,05697

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
pH (25°C; 5 %) 5.0 - 7.0
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 10 ppm
NH4 (Ammonium) Max. 30 ppm
PO4 (Phosphate) Max. 5 ppm
SO4 (Sulphate) Max. 50 ppm
Ba (Barium) Max. 50 ppm
Ca (Calcium) Max. 100 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 10 ppm
Mn (Manganese) Max. 5 ppm
Na (Sodium) Max. 50 ppm
Sr (Strontium) Max. 50 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR