Xylylazo violet I, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]-1-phenol-4-sulphonic acid sodium salt, 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1'-(2-hydroxybenzene-5-sulphonic acid) sodium salt, Xylidyl Blue I

248266-5G
248266-5GEA 102 GBP
248266-5G
Xylylazo violet I, Sigma-Aldrich®
Xylylazo Violet I

Xylidyl blue I, (4-Hydroxy-3-[2-hydroxy-3-(2,4-dimethylphenylaminocarbonyl)-1-naphthylazobenzenesulfonic acid sodium salt; Magonsulfonate)

Formula: C₂₅H₂₁N₃O₆S
MW: 513.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003890
CAS Number: 14936-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9573c93288d53d81e1df2db76d649b0fef41e44&vid=cf062a4132233df80fff336efbfd228a2a08e91f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR