VWR®, Laboratory Jacks
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
245-0004EA 208 GBP
245-0004 245-0005 245-0006 245-0007 245-0008 245-0009
VWR®, Laboratory Jacks
Laboratory Jacks
Anodised aluminium. With M10 threaded hole in the middle of the top plate for attaching retort rods (starting from item number 245-0006).
  • Smooth adjustment of working height
  • Extreme stability
  • Exceptionally sturdy
  • Corrosion resistant and non susceptible to a wide range of chemicals