Reactive blue 4 35% (dye content), Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

244813-25G
244813-25GEA 64.4 GBP
244813-25G
Reactive blue 4 35% (dye content), Sigma-Aldrich®
Reactive blue 4

Reactive Blue 4, Dye content 35 % (1-Amino-4-[3-(4,6-dichlorotriazin-2-ylamino)-4-sulfophenylamino]anthraquinone-2-sulfonic acid; Procion(R) blue MX-R)

Formula: C₂₃H₁₄Cl₂N₆O₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00001218
CAS Number: 13324-20-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5440c905d17f0eabc06af0cdc829dac3e9e4cda3&vid=a5f048f38cb6c19270de8ad925c6a86a3eaf51fa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR