Florisil® 60-100 mesh for chromatography

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

24278.182EA 73.8 GBP
24278.182 24278.295 24278.230 24278.440
Florisil® 60-100 mesh for chromatography
Magnesium silicate
Formula: MgO₃Si
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

Identification Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR