Iron(III) chloride, anhydrous, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Iron trichloride, Ferric chloride

24216.268EA 45.6 GBP
24216.268 24216.361 24216.460
Iron(III) chloride, anhydrous, GPR RECTAPUR®
Iron (III) chloride
Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 315
Melting Pt: 260
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 1773
ADR: 8 III
Merck Index: 13,04048

Order Now

Specification Test Results

Appearance Dark green to black, cryst. powder (NK)
Assay Min. 98,5%
Insoluble matter Max. 0,2%
Arsenic (As) Max. 0.001%
Chromium (Cr) Max. 0,05%
Copper (Cu) Max. 0,01%
Manganese (Mn) Max. 0,2%
Nickel (Ni) Max. 0,03%
Lead (Pb) Max. 0.001%
Zinc (Zn) Max. 0,04%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR