Stand bases
Supplier: R L ENTERPRISES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3107/1 3107/11 3107/2 3107/21 3107/3 3107/31 3107/4 3107/41
241-0173EA 72.2 GBP
241-0173 241-0174 241-0176 241-0177 241-0179 241-7869 241-0182 241-0183
Stand bases
Retort Stands
Rectangular retort bases with M10 threaded hole in centre of short side.
  • Steel, with acrylic finish
  • Acid resistant surface
  • Non-slip rubber feet for a firm footing
Ordering information: Supplied without rods which need to be ordered separately.