Silica gel, Pore size: 60 Å 0.060-0.200 mm for chromatography

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

240370010 240370050 240370300
240370010.EA 61.1 GBP
240370010. 240370050. 240370300.
Silica gel, Pore size: 60 Å 0.060-0.200 mm for chromatography
Silica gel
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9

Order Now

Specification Test Results

Appearance White Powder
Particle size ≤5 % (Oversize)
Specific gravity 0.46 to 0.55 kg/l
pH 6.5 to 7.5 (5 % suspension in water)
Residual water 2 to 4 % (160°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR