Tetraethyl orthosilicate (TEOS) ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

24004.290EA 87 GBP
24004.290
Tetraethyl orthosilicate (TEOS) ≥99%, GPR RECTAPUR®
Tetraethyl orthosilicate (TEOS)
Formula: Si(OC₂H₅)₄
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: –85,5 °C
Density: 0.933 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00009062
CAS Number: 78-10-4
EINECS: 201-083-8
UN: 1292
ADR: 3,III
Merck Index: 13,03882

Order Now

Specification Test Results

Assay Min. 99 %
Acidity Max. 0.002 meq/g
Density (20/4) 0.930 - 0.940
Cl (Chloride) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR