2-(Cyclohexylamino)ethanesulphonic acid (CHES), Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N-Cyclohexyltaurine, CHES

239779-100
239779-100EA 146 GBP
239779-100
2-(Cyclohexylamino)ethanesulphonic acid (CHES), Millipore®
2-(Cyclohexylamino)ethanesulphonic acid (CHES)
Formula: C₈H₁₇NO₃S
MW: 207.29 g/mol
Melting Pt: 320
Storage Temperature: Ambient
MDL Number: MFCD00003835
CAS Number: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR