SafeKey lockout padlocks, aluminium
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
150285 150242 150243 150263 150264 150286 150287 150288 150307 150333 150303 150219 150256 150302 150240 150326 150347 150301 150278 150260 150261 150262 150304 150305 150348 150349 150281 150282 150283 150284 150306 150330 150331 150332 150360 150241 150323 150345 150252 150322 150216 150344 150358 150235 150297 150217 150218 150238 150239 150276 150277 150299 150300 150325
238-0149EA 28.04 GBP
238-0149 238-0133 238-0134 238-0140 238-0141 238-0150 238-0151 238-0152 238-0162 238-0170 238-0158 238-0127 238-0136 238-0157 238-0131 238-0166 238-0173 238-0156 238-0144 238-0137 238-0138 238-0139 238-0159 238-0160 238-0174 238-0175 238-0145 238-0146 238-0147 238-0148 238-0161 238-0167 238-0168 238-0169 238-0177 238-0132 238-0164 238-0172 238-0135 238-0163 238-0124 238-0171 238-0176 238-0128 238-0153 238-0125 238-0126 238-0129 238-0130 238-0142 238-0143 238-0154 238-0155 238-0165
SafeKey lockout padlocks, aluminium
Security Locks
These SafeKey aluminium padlocks are the ultimate locks for indoor and outdoor heavy-duty safety applications.
  • Anodised, corrosion-resistant finish provides extended product life
  • Durable construction for improved tamper resistance
  • Resilient against harsh environments, including food, oils and chemicals
  • Ideal for indoor and outdoor use
  • Engraving options on front, back, left side, right side and/or key
  • Non conductive
Manufactured with solid aluminium body and steel shackle, these padlocks are amazingly lightweight and rugged. They compliment the BradyMore Product Information