Tin(II) chloride dihydrate 98.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Stannous chloride dihydrate

23742.260EA 326 GBP
23742.260 23742.293
Tin(II) chloride dihydrate 98.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Tin (II) chloride dihydrate
Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623
Melting Pt: 246
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay 98.0 - 103.0 %
Identification Passes test
Insolubility in diluted HCl Passes test
NH4 (Ammonium) Max. 20 ppm
SO4 (Sulphate) Max. 20 ppm
As (Arsenic) Max. 1 ppm
Ca (Calcium) Max. 0,005%
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 20 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 100 ppm
Pb (Lead) Max. 50 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR