Ethanol denatured (industrial methylated spirit) ≥99%, GPR RECTAPUR® (denatured with methanol)

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

23684.329EA 79.3 GBP
23684.329 23684.469 23684.360 23684.444
Ethanol denatured (industrial methylated spirit) ≥99%, GPR RECTAPUR® (denatured with methanol)
Ethanol denatured

Delivery information: This product is not available in all countries. Please check with your local VWR office or supplier.

Formula: H₃CCH₂OH
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

ALCH 99.1 - 99.5 %
Density (20/4) 0.792 - 0.794 g/cm3
Colour number (Hazen) Max. 20.0 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR