Hydrogen peroxide 30% Unstabilised, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

23619.297EA 179 GBP
23619.297 23619.264 23619.366
Hydrogen peroxide 30% Unstabilised, AnalaR NORMAPUR® zA
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ~107 °C (1013 hPa)
Melting Pt: <0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8) II
Merck Index: 14,04798

Order Now

Specification Test Results

Assay 30.0-32.0 %
Acidity Max. 0.0002 meq/g
Colouration Max. 10 APHA
Evaporation residue Max. 30 ppm
Heavy metals (as Pb) Max. 1 ppm
Total N (Nitrogen) Max. 4 ppm
Cl (Chloride) Max. 0.5 ppm
PO4 (Phosphate) Max. 2 ppm
SO4 (Sulphate) Max. 2 ppm
As (Arsenic) Max. 0.5 ppm
Cd (Cadmium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR