1,5-Diphenylcarbazide ≥97.0% analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Diphenylcarbazide, sym-Diphenylcarbazide, 1,5-Diphenylcarbohydrazide, 1,5-Diphenylcarbonohydrazide, N'',N'''-Diphenylcarbonohydrazide, sym. Diphenylcarbazide

23550.120EA 37.9 GBP
23550.120 23550.153
1,5-Diphenylcarbazide ≥97.0% analytical reagent
1,5-Diphenylcarbazide
Formula: C₁₃H₁₄N₄O
MW: 242.28 g/mol
Boiling Pt: 454
Melting Pt: 170…172 °C
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 97.0 %
Suited for Cr6+ reagent Passes test
Melting point 171 - 173 °C
Ignition residue (SO4) Max. 0.1 %
Insolubility in methanol Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR