N,N-Dimethylformamide ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

23466.298EA 132 GBP
23466.298 23466.323 23466.550 23466.460 23466.930
N,N-Dimethylformamide ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –60,5 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (DIN 51755) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.8 %
IR Spectrum Passes test
Acidity Max. 0.00025 meq/g
Alkalinity Max. 0.003 meq/g
Boiling point 152 - 155 °C
Colouration Max. 10 APHA
Density (20/4) 0.946 - 0.950
Density (20/20) 0.949 - 0.952
Refractive index (20°C) 1.429 - 1.431
Evaporation residue Max. 10 ppm
Water Max. 0.05 %
Al (Aluminium) Max. 0.5 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.01 ppm
Sr (Strontium) Max. 0.05 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR