Safety knives for left and right-handed users, SECUNORM PROFI
Supplier: MARTOR ARGENTAX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
07152.02
233-2245EA 0 GBP
233-2245
Safety knives for left and right-handed users, SECUNORM PROFI
Knives
Trapezoid blade, aluminium handle.
  • Versions available for left or right-handed users
  • When cutting, the user must remove their thumb from the slider
  • Easy blade replacement by sliding back the inner part, without the need for tools
These classic safety knives have a spring-loaded blade which automatically disappears into the handle if the blade slips off the material being cut and the slider is not held down. They are especially suitable for slitting cardboard boxes and adhesive tape.
TÜV tested
Certifications: TÜV tested