Minor skin graft blade, No. SG3
Supplier: SWANN MORTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2201
233-0053EA 54.4 GBP
233-0053
Minor skin graft blade, No. SG3
Scalpels and Scalpel Assemblies Scalpel Blades
Stainless steel. Originally developed for use with skin grafts, these blades are also suitable for a wide range of other applications.
  • Fits standard No. 3 surgical handle (233-5454)
  • Sterile
  • Individually packed
Ideal for use on the hands and feet of small children given its practical size advantage compared to a standard skin grafting knife. Also suitable for use within Histology departments for the fine sectioning of the pathology blocks.
Packaging: Packed in individual foil packs.