cis-Platin, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride

232120-50
232120-50EA 93.7 GBP
232120-50
cis-Platin, Millipore®
cis-Platin
Formula: H₆Cl₂N₂Pt
Density: 3.738 g/cm³ (20 °C)
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II
Merck Index: 13,02341

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR