Forceps with guide pin
Supplier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
312.130 312.145 312.160 312.200 312.115 312.105
232-2165EA 24.2 GBP
232-2165 232-2166 232-2167 232-0205 232-0078 232-0077
Forceps with guide pin
Forceps
Stainless steel, anti-magnetic. Particularly suitable for staining applications.
  • With curved, extra fine tips
  • Corrosion resistant