VWR®, Clips for Conical Standard Ground Joints
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
232-0053EA 8.5 GBP
232-0053 232-0054 232-0055 232-0056 232-0057 232-0058
VWR®, Clips for Conical Standard Ground Joints
Clamps NS Joint Clamps
PTFE with inner steel spring. Very stable, safe and durable, easy to use.
  • Excellent chemical resistance
  • Temperature resistant from −200 to +280 °C