Double spatulas, micro
Supplier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
373.150 374.150 376.130 376.150 376.185 376.210 374.130 373.210 377.150 374.185 375.150
231-0425EA 42.2 GBP
231-0425 231-1412 231-2321 231-2322 231-2323 231-2324 231-0427 231-0426 231-0430 231-0428 231-0429
Double spatulas, micro
Spatulas
Stainless steel. Anti-magnetic, very flexible micro powder spatula.
  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance