Spoon spatulas
Supplier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
L74/2 L74/4 L74/6 L74/8
231-1342EA 34 GBP
231-1342 231-1344 231-1346 231-1348
Spoon spatulas
Spatulas
Porcelain, glazed white.
  • Broad shape
  • Excellent chemical resistance
  • Heat resistant up to 1000 °C
  • Designed for rough handling