Laboratory spoon
Supplier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
026.200
231-1231EA 14.2 GBP
231-1231
Laboratory spoon
Spoons
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses.
  • Open ended spoon, with flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean