Scoops, FoodScoop
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5371-0100 5371-0200 5371-0350 5371-0500 5371-0250
231-0403EA 47.4 GBP
231-0403 231-0404 231-0406 231-0407 231-0405
Scoops, FoodScoop
Scoops
Stainless steel V2A (1.4301), highly polished. Very stable hand scoop for various applications, particularly suitable for the food industry.
  • Conforms to European food law regulations
  • Robust design
  • Seamless join between scoop and hollow handle so has no crevices and edges