Scoops
Supplier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
758.050 758.075 758.100 758.120 758.150
231-0201EA 24.5 GBP
231-0201 231-0202 231-0203 231-0204 231-0236
Scoops
Scoops
Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Robust design