Tubing connectors, straight
Supplier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
70510 70511 70512 70513 70514 70515
229-0713EA 29.2 GBP
229-0713 229-0714 229-0715 229-0716 229-0717 229-0718
Tubing connectors, straight
Fittings and Connectors
PP, white.
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520
With tapered ends, for connecting tubing with the same diameter.