Cyclohexane 99+% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

22864.AP 22864.K2 22864.K7 22864.M1
22864.APEA 27.7 GBP
22864.AP 22864.K2 22864.K7 22864.M1
Cyclohexane 99+% ACS
Cyclohexane

Purity: 99+% CAS No.: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -18°(-1°F) Physical Form: Liquid Density: 0.779 Refractive Index: 1.4260 Odour: Pungent Melting Point: 6.5° Boiling Point: 80-81° Beilstein Reference: 1900225 Merck Reference: 14,2723 UN No.: 1145 MDL No.: MFCD00003814

Formula: C₆H₁₂
Boiling Pt: 80…81 °C
Melting Pt: 6.5 °C
Density: 0.779
Flash Pt: –18 °C (–1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 14,2723
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR