Tubing, PTFE, BOLA
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S1810-02 S1810-04 S1810-05 S1810-06 S1810-08 S1810-09 S1810-10 S1810-12 S1810-14 S1810-16 S1810-18 S1810-19 S1810-20 S1810-22 S1810-24 S1810-28 S1810-30 S1810-32 S1810-33 S1810-34 S1810-36 S1810-37 S1810-38 S1810-40 S1810-42 S1810-44 S1810-46 S1810-48 S1810-50 S1810-52 S1810-54 S1810-56 S1810-58 S1810-60 S1810-62 S1810-64 S1810-66 S1810-68 S1810-70 S1810-72 S1810-74 S1810-88 S1810-84 S1810-21 S1810-26 S1810-63 S1810-78 S1810-01 S1810-61 S1810-23 S1810-39 S1810-43
228-0732EA 6 GBP
228-0732 228-0733 228-0734 228-0735 228-4108 228-0736 228-0737 228-0738 228-4118 228-1496 228-4119 228-0739 228-4124 228-4125 228-0741 228-0743 228-4134 228-4135 228-0744 228-4138 228-0745 228-1499 228-4139 228-0746 228-0747 228-0748 228-4140 228-0749 228-0750 228-0751 228-1529 228-1530 228-0752 228-0753 228-0754 228-4168 228-1533 228-4109 228-1534 228-0755 228-0756 228-1536 228-1198 228-1497 228-0742 228-1532 228-1535 228-1495 228-1531 228-1498 BOHLS1810-39 BOHLS1810-43
Tubing, PTFE, BOLA
Tubing Transfer Tubing
PTFE, white, translucent.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible
This tubing is characterised by its non stick surface and excellent dielectric properties.
Temperature resistance: −200 to +260 °C