Tubing, silicone
Supplier: DEUTSCH NEUMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3102026 3101018 3102545 3100405 3101015/25 3100912 3101016 3100525B 3101523A 3101535A 3101216B 3101218 3101319 3101420 3101521 3101621 3100510/25 3101115 3100804 3100409 3102534 3100205/25M 3100305/25 3100307/25 3100407/25 3100508/25 3100609/25 3100812/25 3100103/25 3100204/25 3100104/25 3100509/25 3100306/25 3100506 3101504 3100106 3100814/25 3100118B 3100206 3100226B 3100406 3100408 3100504 3100507 3100610 3100710B 3100711 3100810 3100811 3100815 3100913 3101014 3100608 3100709 3101013/25 3100228/10 3102533 3101503 3100228 3101013 3101824 3100338
228-1571EA 951 GBP
228-1571 228-1573 228-1579 228-1580 228-3569 228-3571 228-3574 228-3861 228-3864 228-3865 228-3888 228-3889 228-3890 228-3891 228-3892 228-3893 228-3894 228-3896 DENE3100804 DENE3100409 DENE3102534 DENE3100205/25M DENE3100305/25 DENE3100307/25 DENE3100407/25 DENE3100508/25 DENE3100609/25 DENE3100812/25 DENE3100103/25 DENE3100204/25 DENE3100104/25 DENE3100509/25 DENE3100306/25 DENE3100506 DENE3101504 DENE3100106 DENE3100814/25 DENE3100118B DENE3100206 DENE3100226B DENE3100406 DENE3100408 DENE3100504 DENE3100507 DENE3100610 DENE3100710B DENE3100711 DENE3100810 DENE3100811 DENE3100815 DENE3100913 DENE3101014 DENE3100608 DENE3100709 DENE3101013/25 DENE3100228/10 DENE3102533 DENE3101503 DENE3100228 DENE3101013 DENE3101824 DENE3100338
Tubing, silicone
Tubing Transfer Tubing
Silicone, translucent.
  • Good chemical resistance
For laboratory application.
Temperature resistance: -60 to +200 °C
Hardness (Shore durometer A): 60
Certifications: BFR XV, FDA CFR 21 § 177.2600, EP -Europ. Pharmacopoeia, Test for biocompatibility according to ISO 10993-1