Chloroform ≥99% stabilised, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Stabilisation: Stabilised with 2-methyl-2-butene (iso-amylene) 0.002 %

22707.295EA 43.2 GBP
22707.295 22707.364 22707.466 22707.320 22707.550
Chloroform ≥99% stabilised, GPR RECTAPUR®
Chloroform
Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 01-2119486657-20
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Boiling point 59.5 - 61.5 °C
Density (20/4) 1.478 - 1.489
Evaporation residue Max. 10 ppm
Free chlorine Max. 10 ppm
Heavy metals (as Pb) Max. 10 ppm
Water Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR