Strontium dodecairon nonadecaoxide ≥99.5% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

22569.06 22569.14
22569.06EA 50.4 GBP
22569.06 22569.14
Strontium dodecairon nonadecaoxide ≥99.5% (metals basis)
Strontium dodecairon nonadecaoxide

Purity: 99.5% CAS No.: 12023-91-5 Molecular Formula: SrFe12O19 Formula Weight: 1061.77 Physical Form: -325 Mesh Powder Melting Point: 450° MDL No.: MFCD00075632

Formula: SrFe₁₂O₁₉
Melting Pt: >450 °C
Storage Temperature: Ambient
MDL Number: MFCD00075632
CAS Number: 12023-91-5
EINECS: 234-685-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR