Carborundum, TECHNICAL 1,680 mm

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

22488.297EA 234 GBP
22488.297
Carborundum, TECHNICAL 1,680 mm
Carborundum
Formula: SiC
Boiling Pt: < 2700 °C (1013 hPa)
Melting Pt: 2700 °C
Density: 3.23 g/cm³ (20 °C)
CAS Number: 409-21-2
EINECS: 206-991-8
Merck Index: 12,08636

Order Now

Specification Test Results

Identification Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR