Calcium hydroxide ≥96%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

22355.298EA 42.9 GBP
22355.298 22355.367
Calcium hydroxide ≥96%, GPR RECTAPUR®
Calcium hydroxide
Formula: Ca(OH)₂
Melting Pt: 550 °C
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III
Merck Index: 13,01675

Order Now

Specification Test Results

Assay (at filling) Min. 96 %
Heavy metals (as Pb) Max. 50 ppm
Insolubility in hydrochloric acid Max. 0.1 %
Not precipitated by (COONH4)2 (as SO4) Max. 2.5 %
Cl (Chloride) Max. 100 ppm
CO3 (as CaCO3) Max. 3 %
SO4 (Sulphate) Max. 0.2 %
Fe (Iron) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR