Hydrofluoric acid 48 - 51% in water for analysis

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

223330250 223335000 223330025
223330250.EA 25.5 GBP
223330250. 223335000. 223330025.
Hydrofluoric acid 48 - 51% in water for analysis
Hydrofluoric acid
Formula: HF
MW: 20.01 g/mol
Melting Pt: –35 °C
Density: 1.15 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Residue after ignition ≤0.0005 %
Titration with NaOH 48 to 51 %
Chloride (Cl) ≤5 ppm
Sulfates ≤5 ppm (SO4)
Phosphates ≤1 ppm (PO4)
Arsenic (As) ≤0.05 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤0.05 ppm
Fluorosilicic acid ≤0.01 % (H2SiF6)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR