Calcium chloride hexahydrate ≥97%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

22311.297EA 172 GBP
22311.297 22311.366
Calcium chloride hexahydrate ≥97%, GPR RECTAPUR®
Calcium chloride hexahydrate
Formula: CaCl₂·6H₂O
Melting Pt: 29.54 °C
Density: 1.71 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149616
CAS Number: 7774-34-7
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Assay Min. 97 %
Heavy metals (as Pb) Max. 20 ppm
SO4 (Sulphate) Max. 0.05 %
Fe (Iron) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR